CAS RN 128-20-1
CAS RN 128-20-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Tocris Bioscience™ Pregnanolone
Postitive allosteric modulator of GABAA receptors