CAS RN 132499-82-2
CAS RN 132499-82-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Malonic acid disodium salt hydrate, 99%
CAS: 132499-82-2 Formula molecolare: C3H2Na2O4 Molecular Weight (g/mol): 148.03 Numero MDL: MFCD00149114 InChI Key: PRWXGRGLHYDWPS-UHFFFAOYSA-L IUPAC Name: disodium propanedioate SMILES: [Na+].[Na+].[O-]C(=O)CC([O-])=O