CAS RN 1620278-72-9
CAS RN 1620278-72-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
TLR7/8 agonist 1 dihydrochloride, MedChemExpress
MedChemExpress TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline.