CAS RN 1781835-20-8
IUPAC Nome:
2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide hydrochloride
Sinonimi:
AdipoRon hydrochloride
Peso molecolare (g/mol):
464.99
Formula molecolare:
C27H29ClN2O3
InChi Key:
TZVJQEGKRLDTHQ-UHFFFAOYSA-N
SMILES:
Cl.O=C(COC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)NC1CCN(CC2=CC=CC=C2)CC1
1
–
2
di
2
risultati
2
R&D Systems™ AdipoRon hydrochloride
A Tocris product, part of R&D Systems. Orally active AdipoR1 and AdipoR2 agonist