CAS RN 1965-09-9
IUPAC Nome:
4-(4-hydroxyphenoxy)phenol
Sinonimi:
phenol, 4,4'-oxybis-
Peso molecolare (g/mol):
202.21
Formula molecolare:
C12H10O3
InChi Key:
NZGQHKSLKRFZFL-UHFFFAOYSA-N
SMILES:
OC1=CC=C(OC2=CC=C(O)C=C2)C=C1
1
–
4
di
4
risultati
1
4,4'-Dihydroxydiphenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,4'-Dihydroxydiphenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,4'-Dihydroxydiphenyl Ether, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.