CAS RN 342777-54-2
CAS RN 342777-54-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
Tocris Bioscience™ LM22B 10
TrkB and TrkC agonist; brain penetrant
2
LM22B-10, MedChemExpress
MedChemExpress LM22B-10 is an activator of TrkB/TrkC neurotrophin receptor, and can induce TrkB, TrkC, AKT and ERK activation in vitro and in vivo.