CAS RN 3605-01-4
CAS RN 3605-01-4
IUPAC Nome:
2-{4-[(2H-1,3-benzodioxol-5-yl)methyl]piperazin-1-yl}pyrimidine; methanesulfonic acid
Sinonimi:
piribedil
Peso molecolare (g/mol):
394.45
Formula molecolare:
C17H22N4O5S
InChi Key:
DEZKFCIOSKCCHK-UHFFFAOYSA-N
SMILES:
CS(O)(=O)=O.C(N1CCN(CC1)C1=NC=CC=N1)C1=CC=C2OCOC2=C1
1
–
1
di
1
risultati
1
Piribedil, MedChemExpress
MedChemExpress Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.