CAS RN 52771-21-8
IUPAC Nome:
3-(trifluoromethoxy)benzaldehyde
Sinonimi:
3-(trifluoromethoxy)benzaldehyde
Peso molecolare (g/mol):
190.12
Formula molecolare:
C8H5F3O2
InChi Key:
FQEVHRCPXFKJHF-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC=CC(C=O)=C1
1
–
2
di
2
risultati
1
3-(Trifluoromethoxy)benzaldehyde, 95%, Thermo Scientific™
CAS: 52771-21-8 Formula molecolare: C8H5F3O2 Molecular Weight (g/mol): 190.12 Numero MDL: MFCD00042406 InChI Key: FQEVHRCPXFKJHF-UHFFFAOYSA-N Sinonimo: 3-trifluoromethoxy benzaldehyde,m-trifluoromethoxy benzaldehyde,m-trifluoromethoxybenzaldehyde,pubchem1470,acmc-209l1m,3trifluoromethoxybenzaldehyde,3-trifluormethoxy-benzaldehyde,ksc274m0j,3-trifluoromethoxy-benzaldehyde,timtec-bb sbb006601 PubChem CID: 605139 IUPAC Name: 3-(trifluoromethoxy)benzaldehyde SMILES: FC(F)(F)OC1=CC=CC(C=O)=C1