CAS RN 5878-19-3
CAS RN 5878-19-3
IUPAC Nome:
1-methoxypropan-2-one
Sinonimi:
1-methoxy-2-propanone
Peso molecolare (g/mol):
88.11
Formula molecolare:
C4H8O2
InChi Key:
CUZLJOLBIRPEFB-UHFFFAOYSA-N
SMILES:
COCC(C)=O
No Raffinamenti
1
–
0
di
0
risultati
1
Methoxyacetone, 96%
CAS: 5878-19-3 Formula molecolare: C4H8O2 Molecular Weight (g/mol): 88.11 Numero MDL: MFCD00008768 InChI Key: CUZLJOLBIRPEFB-UHFFFAOYSA-N Sinonimo: methoxyacetone,methoxy-2-propanone,2-propanone, 1-methoxy,1-methoxy-2-propanone,1-methoxyacetone,methoxyaceton,methoxymethyl methyl ketone,methoxy acetone,ch3coch2och3 PubChem CID: 22172 IUPAC Name: 1-methoxypropan-2-one SMILES: COCC(C)=O