CAS RN 61341-86-4
IUPAC Nome:
2,3-dihydro-1H-inden-1-amine
Sinonimi:
indanamin
Peso molecolare (g/mol):
133.19
Formula molecolare:
C9H11N
InChi Key:
XJEVHMGJSYVQBQ-UHFFFAOYNA-N
SMILES:
NC1CCC2=CC=CC=C12
1
–
3
di
3
risultati
2
(S)-1-Aminoindane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.