CAS RN 66611-27-6
CAS RN 66611-27-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Tocris Bioscience™ RU 24969 hemisuccinate
5-HT1A and 5-HT1B agonist
2
RU 24969 Succinate (2:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
RU 24969 Succinate (2:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.