CAS RN 72581-71-6
CAS RN 72581-71-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Isosilybin, MedChemExpress
MedChemExpress Isosilybin (Isosilybinin) is a flavonoid from milk thistle; inhibits CYP3A4 induction with an IC50 of 74 μM.