CAS RN 757949-98-7
CAS RN 757949-98-7
IUPAC Nome:
2-[2-(3-fluorophenyl)ethynyl]-4,6-dimethylpyridin-3-amine hydrochloride
Sinonimi:
raseglurant hydrochloride
Peso molecolare (g/mol):
276.74
Formula molecolare:
C15H14ClFN2
InChi Key:
BDXXMGYKLQZXOU-UHFFFAOYSA-N
SMILES:
Cl.CC1=NC(C#CC2=CC(F)=CC=C2)=C(N)C(C)=C1
1
–
1
di
1
risultati
1
Tocris Bioscience™ ADX 10059 hydrochloride
CAS: 757949-98-7 Formula molecolare: C15H14ClFN2