CAS RN 85-68-7
IUPAC Nome:
1-benzyl 2-butyl benzene-1,2-dicarboxylate
Sinonimi:
benzyl butyl phthalate butylbenzyl phthalate unimoll BB butyl benzyl phthalate (substance) benzyl butyl phthalate (bbp) BBP sicol 160 sicol caswell no. 125G ccris 104
Peso molecolare (g/mol):
312.37
Formula molecolare:
C19H20O4
InChi Key:
IRIAEXORFWYRCZ-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
1
–
5
di
5
risultati
1
Benzyl Butyl Phthalate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Phthalic acid, benzylbutyl ester, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
Phthalic acid, benzylbutyl ester 1000 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality