CAS RN 88150-45-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
Ethyl 4-(2-Azidoethoxy)-3-oxobutanoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Ethyl (2-Azidoethoxy)acetoacetate, TRC
CAS: 88150-45-2 Formula molecolare: C8 H13 N3 O4 Molecular Weight (g/mol): 215.21 Sinonimo: Butanoic acid, 4-(2-azidoethoxy)-3-oxo-, ethyl ester,4-(2-Azidoethoxy)-3-oxobutanoic acid ethyl ester IUPAC Name: ethyl 4-(2-azidoethoxy)-3-oxobutanoate SMILES: CCOC(=O)CC(=O)COCCN=[N+]=[N-]