CAS RN 94134-94-8
CAS RN 94134-94-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
5-Methylpyrrolidin-3-ol, TRC
CAS: 94134-94-8 Formula molecolare: C5 H11 N O Molecular Weight (g/mol): 101.15 IUPAC Name: 5-methylpyrrolidin-3-ol SMILES: CC1CC(O)CN1