CAS RN 945966-46-1
CAS RN 945966-46-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Apararenone, MedChemExpress
MedChemExpress Apararenone (MT-3995) is a novel non-steroidal mineralocorticoid receptor antagonists under development for the treatment of diabetic nephropathies and non-alcoholic steatohepatitis.