CAS RN 947-91-1
CAS RN 947-91-1
IUPAC Nome:
2,2-diphenylacetaldehyde
Sinonimi:
benzeneacetaldehyde, α-phenyl-
Peso molecolare (g/mol):
196.25
Formula molecolare:
C14H12O
InChi Key:
HLLGFGBLKOIZOM-UHFFFAOYSA-N
SMILES:
O=CC(C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
di
2
risultati
1
Diphenylacetaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Diphenylacetaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.