CAS RN 949142-50-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
MedChemExpress Obinutuzumab
MedChemExpress Obinutuzumab (GA101) a novel glycoengineered Type II CD20 humanized IgG1 monoclonal antibody in development for non-Hodgkin lymphoma.
2
MedChemExpress Obinutuzumab
MedChemExpress Obinutuzumab (GA101) a novel glycoengineered Type II CD20 humanized IgG1 monoclonal antibody in development for non-Hodgkin lymphoma.