CAS RN 957054-30-7
CAS RN 957054-30-7
IUPAC Nome:
4-{6-[(4-methanesulfonylpiperazin-1-yl)methyl]-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl}-1H-indazole
Sinonimi:
4-{6-[(4-methanesulfonylpiperazin-1-yl)methyl]-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl}-1H-indazole
Peso molecolare (g/mol):
513.64
Formula molecolare:
C23H27N7O3S2
InChi Key:
LHNIIDJUOCFXAP-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)N1CCN(CC2=CC3=NC(=NC(N4CCOCC4)=C3S2)C2=C3C=NNC3=CC=C2)CC1
1
–
5
di
5
risultati
2
GDC-0941, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
GDC-0941, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
GDC-0941, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Pictilisib, MedChemExpress
MedChemExpress Pictilisib (GDC-0941) is a potent inhibitor of PI3Kα/δ with an IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).