CAS RN 97-17-6
CAS RN 97-17-6
IUPAC Nome:
O-2,4-dichlorophenyl O,O-diethyl phosphorothioate
Sinonimi:
dichlofenthion
Peso molecolare (g/mol):
315.14
Formula molecolare:
C10H13Cl2O3PS
InChi Key:
WGOWCPGHOCIHBW-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)OC1=CC=C(Cl)C=C1Cl
1
–
4
di
4
risultati
1
Dichlofenthion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dichlofenthion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dichlofenthion, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.