CAS RN 97928-18-2
CAS RN 97928-18-2
IUPAC Nome:
1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
Sinonimi:
1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
Peso molecolare (g/mol):
303.45
Formula molecolare:
C19H29NO2
InChi Key:
HUFTWIJFTZKVMR-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN1CCC(O)CC1
1
–
2
di
2
risultati
1
1-[3-(4-tert-Butylbenzoyl)propyl]-4-hydroxypiperidine, TRC
CAS: 97928-18-2 Formula molecolare: C19 H29 N O2 Molecular Weight (g/mol): 303.44 Sinonimo: 1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one,Ebastine Imp. D (EP) IUPAC Name: 1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one SMILES: CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(O)CC2
2
1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.