CAS RN 98475-07-1
CAS RN 98475-07-1
IUPAC Nome:
methyl 2-(bromomethyl)-3-nitrobenzoate
Sinonimi:
methyl 2-(bromomethyl)-3-nitrobenzoate
Peso molecolare (g/mol):
274.07
Formula molecolare:
C9H8BrNO4
InChi Key:
FCGIVHSBEKGQMZ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(CBr)C(=CC=C1)[N+]([O-])=O
1
–
4
di
4
risultati
1
2-(Bromomethyl)-3-nitrobenzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl 2-(Bromomethyl)-3-nitrobenzoate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
2-(Bromomethyl)-3-nitrobenzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-(Bromomethyl)-3-nitrobenzoic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.