Cell Culture Flasks
Cell culture flasks are essential for consistent growth and expansion of adherent and suspension cell lines. In this category, you’ll find a broad range of sterile and cell-culture treated options, including vented and filtered closures to support gas exchange and contamination control. Explore trusted ranges from Corning, Thermo Scientific Nunc, Thermo Scientific, and Fisherbrand to match your workflow—from routine maintenance to scale-up.
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks
These polystyrene flasks receive the proprietary Nunclon Delta surface treatment to maximize adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface for pipettes or cell scrapers.
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks
Corning™ Polycarbonate Erlenmeyer Flasks
Manufactured from optically clear polycarbonate for optimum visibility and durability. Ideal for shaker cultures applications.
Corning™ Polycarbonate Erlenmeyer Flasks
Corning™ U-Shaped Cell Culture Flasks
Designed to improve usability while maintaining the same environment for cell growth as previous designs.
Corning™ U-Shaped Cell Culture Flasks
Complete Your Order - Great Deals
Cell Culture Flask Accessories
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ System Accessories.
Cell Culture Flask Accessories
Serological Pipets
Our range of serological pipets are used for transferring liquids and mixing chemical solutions or cell suspensions with the help of a pipette dispenser.
Serological Pipets
Centrifuge Tubes
Our range of glass and plastic centrifuge tubes are available in a variety of volume capacities, materials, configurations, RCF ratings, and closure types. They are available both sterile or non-sterile and may include graduations indicating volume.
Centrifuge Tubes
FAQ
Vented caps support gas exchange, while filtered caps add a barrier that helps reduce contamination risk during handling. For routine CO2 incubator work, many labs choose filtered options such as Thermo Scientific Nunc EasYFlasks, while vented Corning-style designs can suit robust workflows.
Choose the flask size based on your target cell yield and split ratio. Smaller areas (25–75 cm²) suit maintenance and early expansion; 175 cm² supports scale-up with fewer vessels. Corning and Thermo Scientific ranges offer consistent footprints to keep seeding densities repeatable.
For adherent lines, select cell-culture treated flasks to improve attachment and growth consistency. Treated surfaces (often labeled TC-treated or Nunclon Delta) help reduce variability across passages. Corning TC-treated flasks and Thermo Scientific Nunc options are widely used for routine adherent culture.
Work in a clean biosafety cabinet, limit cap-off time, and avoid touching neck threads or inner surfaces. Use sterile techniques when adding media and cells, and store unopened sterile packs properly. Filtered closures (e.g., Thermo Scientific Nunc) can provide extra protection during routine handling steps.
Cell culture flasks are used for seeding, maintenance, passaging, and expansion before downstream assays, imaging, or bioprocess steps. Many labs start in 25–75 cm² formats and move to 175–225 cm² as cell numbers increase. Corning and Thermo Scientific portfolios support each stage with matching formats.
Use the same flask type (surface treatment and closure style), standardize seeding density, and keep incubation conditions consistent. Selecting trusted brands like Corning, Thermo Scientific, and Thermo Scientific Nunc helps reduce lot-to-lot variability, while choosing sterile, ready-to-use formats minimizes handling differences.
Top Brands