Improve Lab Productivity with Column Modernization
Have you changed LC column dimensions? Do you have existing columns that are not available anymore, or suppliers who are no longer trading? Are you interested in modernizing existing methods to increase lab productivity?
Reduce Time and Solvent Usage With Newer Column Technology
Major steps have been taken by the US Pharmacopeia to allow labs to improve upon their methods, effectively saving time and money as well as reducing solvent consumption. As of 01 December 2022, the US Pharmacopeia will allow for changes to particle size, length, and inner diameter for both isocratic and gradient methods in order to improve the original methods to new standards (Table 1). The EU Pharmacopoeia and Japanese Pharmacopoeia have followed suit with their newest editions, which have been implemented since 01 January 2023.
Variable
Isocratic Separation
Gradient Separation
Stationary Phase
Mobile Phase pH
Particle Size/Column Length
Injection Volume
Flow Rate
Column Temperature
No change in the physio-chemical characteristics of the stationary phase (same L category)
±0.2 pH units (unless otherwise described)
Per constant L/dp or N: −25% to +50%
Optional/Flexible
An additional change in flow rate of ±50% is permitted
After the flow rate calculation, change in flow rate is not permitted
±10°C
±5°C
Table 1. Changes to the US and EU Pharmacopoeia
The following (Table 2) is a listing of suitable Thermo Scientific™ chromatography columns for LC chromatography according to USP 621, which can also be applicable for EU Pharmacopeia.
L Category
Description
Thermo Scientific™ Columns
L1
Octadecyl silane chemically bonded to porous or non-porous silica or ceramic micro-particles (1.5 to 10μm in diameter) or a monolithic rod
Acclaim™ 120 C18
Acclaim™ RSLC C18
Accucore™ C18
Accucore™ aQ
Accucore™ 150-C18
Accucore™ XL C18
AQUASIL™ C18
Hyperprep™ HS C18
Hypersil™ 100 C18
Hypersil™ BDS C18
Hypersil™ GOLD
Hypersil™ GOLD aQ
Hypersil™ ODS
Hypersil™ ODS-2
Syncronis™ C18
Syncronis™ aQ
Table 2. HPLC Column Selection by USP Specification
Click here to view more USP codes, descriptions, and recommended phases.
LC Columns
Content provided by:
Thermo Scientific logo
Reference