missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine, 96%, Thermo Scientific Chemicals

Catalog Number p-7031838
Click to view available options
Quantità:
250 mg
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
84162-82-3
C14H15F2NO2
267.276
MFCD00179440
UGPWQEIGJWVJDR-UHFFFAOYSA-N
4-2',4'-difluorobenzoyl-1-acetylpiperidine, 1-4-2,4-difluorobenzoyl piperidin-1-yl ethanone, 1-acetylpiperidin-4-yl 2,4-difluorophenyl methanone, 1-acetyl-4-2,4-difluorobenzoyl piperidine, 1-4-2,4-difluoro-benzoyl-piperidin-1-yl-ethanone, 1-acetyl-4-2',4'-difluorobenzoyl piperidine, 1-4-2,4-difluorobenzoyl-1-piperidinyl-1-ethanone, 1-4-2.4-difluoro-benzoyl-piperidin-1-yl-ethanone, maybridge3_006593
2740707
1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethanone
CC(=O)N1CCC(CC1)C(=O)C2=C(C=C(C=C2)F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
84162-82-3
C14H15F2NO2
267.276
MFCD00179440
UGPWQEIGJWVJDR-UHFFFAOYSA-N
4-2',4'-difluorobenzoyl-1-acetylpiperidine, 1-4-2,4-difluorobenzoyl piperidin-1-yl ethanone, 1-acetylpiperidin-4-yl 2,4-difluorophenyl methanone, 1-acetyl-4-2,4-difluorobenzoyl piperidine, 1-4-2,4-difluoro-benzoyl-piperidin-1-yl-ethanone, 1-acetyl-4-2',4'-difluorobenzoyl piperidine, 1-4-2,4-difluorobenzoyl-1-piperidinyl-1-ethanone, 1-4-2.4-difluoro-benzoyl-piperidin-1-yl-ethanone, maybridge3_006593
2740707
1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethanone
CC(=O)N1CCC(CC1)C(=O)C2=C(C=C(C=C2)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1-Acetyl-4-(2',4'-difluorobenzoyl)piperidine
Punto di fusione 97°C to 98°C
Quantità 250 mg
Peso formulazione 267.28
Percent Purity 96%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'