Learn More
Hamilton™ 1000 Series Gastight Instrument Syringes
With a precision-machined PTFE plunger tip which creates a leak-free seal.
Marca: Hamilton™ 208643
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Tip wipes the interior of the syringe barrel free of sample
- Useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze
- Gas sterilizable
Specifica
2.5 mL |