Learn More
Fisherbrand™ 14-Tooth Microplate Format Combs for Horizontal Midigel Systems
14-Tooth Microplate Format Combs for Horizontal Midigel Systems
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Accessory for use with Horizontal Midigel Systems
Specifica
Owl™ A1 Large Gel Electrophoresis System | |
Microplate | |
0.15 cm | |
0.28 in. |
Standard Comb | |
14 | |
50 μL | |
0.72 cm |