missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol, 97%, Thermo Scientific Chemicals

Catalog Number 11383247
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7594-49-2
C4HCl3F6O
285.391
MFCD00236021
AMCTWDASUQVRCQ-UHFFFAOYSA-N
1,1-bis trifluoromethyl-2,2,2-trichloroethanol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl propan-2-ol, 2-trichloromethylhexafluoro-2-propanol, 2,2,2-trichloro-1,1-bis trifluoromethyl ethanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl-2-propanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl propan-2-ol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl-2-propanol, 1,1,1-tris chloranyl-3,3,3-tris fluoranyl-2-trifluoromethyl propan-2-ol
2773244
1,1,1-trichloro-3,3,3-trifluoro-2-(trifluoromethyl)propan-2-ol
C(C(F)(F)F)(C(F)(F)F)(C(Cl)(Cl)Cl)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7594-49-2
C4HCl3F6O
285.391
MFCD00236021
AMCTWDASUQVRCQ-UHFFFAOYSA-N
1,1-bis trifluoromethyl-2,2,2-trichloroethanol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl propan-2-ol, 2-trichloromethylhexafluoro-2-propanol, 2,2,2-trichloro-1,1-bis trifluoromethyl ethanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl-2-propanol, 1,1,1,3,3,3-hexafluoro-2-trichloromethyl propan-2-ol, 1,1,1-trichloro-3,3,3-trifluoro-2-trifluoromethyl-2-propanol, 1,1,1-tris chloranyl-3,3,3-tris fluoranyl-2-trifluoromethyl propan-2-ol
2773244
1,1,1-trichloro-3,3,3-trifluoro-2-(trifluoromethyl)propan-2-ol
C(C(F)(F)F)(C(F)(F)F)(C(Cl)(Cl)Cl)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
Punti di ebollizione 139°C
Indice di rifrazione 1.389
Quantità 1 g
Beilstein 1841641
Peso formulazione 285.4
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'