missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-cyano-5-phenylpyridine, 98%, Thermo Scientific™

Catalog Number 11395609
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
10177-10-3
C12H7ClN2
214.652
MFCD00231610
DBOYDCQWVLJOJG-UHFFFAOYSA-N
2-chloro-5-phenylnicotinonitrile, 2-chloro-3-cyano-5-phenylpyridine, 2-chloro-5-phenyl-pyridine-3-carbonitrile, 3-pyridinecarbonitrile, 2-chloro-5-phenyl, acmc-20anyy, chlorophenylnicotinonitrile
2764043
2-chloro-5-phenylpyridine-3-carbonitrile
C1=CC=C(C=C1)C2=CN=C(C(=C2)C#N)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
10177-10-3
C12H7ClN2
214.652
MFCD00231610
DBOYDCQWVLJOJG-UHFFFAOYSA-N
2-chloro-5-phenylnicotinonitrile, 2-chloro-3-cyano-5-phenylpyridine, 2-chloro-5-phenyl-pyridine-3-carbonitrile, 3-pyridinecarbonitrile, 2-chloro-5-phenyl, acmc-20anyy, chlorophenylnicotinonitrile
2764043
2-chloro-5-phenylpyridine-3-carbonitrile
C1=CC=C(C=C1)C2=CN=C(C(=C2)C#N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Chloro-3-cyano-5-phenylpyridine
Punto di fusione 143°C to 147°C
Quantità 250 mg
Peso formulazione 214.66
Percent Purity 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'