missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyano-4-methylpyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantità:
250 mg
1 g
5 g
Descrizione
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifica
Materiale o nome chimico | 2-Cyano-4-methylpyridine |
Punto di fusione | 85°C to 88°C |
Punti di ebollizione | 145°C to 148°C (38 mmHg) |
Odore | Characteristic |
Quantità | 1 g |
Beilstein | 110753 |
Informazioni di solubilità | Slightly soluble in water. |
Peso formulazione | 118.14 |
Percent Purity | 98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'