Learn More
2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog Number 11493867
Click to view available options
Quantità:
5 g
25 g
38985-64-7
C7H4FNS
153.17
MFCD00004800
OAGDRIUTLPDSMJ-UHFFFAOYSA-N
2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546
97520
1-fluoro-2-isothiocyanatobenzene
FC1=CC=CC=C1N=C=S
38985-64-7
C7H4FNS
153.17
MFCD00004800
OAGDRIUTLPDSMJ-UHFFFAOYSA-N
2-fluorophenyl isothiocyanate, o-fluorophenyl isothiocyanate, benzene, 1-fluoro-2-isothiocyanato, 2-fluorophenylisothiocyanate, 1-fluoro-2-isothiocyanato-benzene, isothiocyanic acid 2-fluorophenyl ester, benzene,1-fluoro-2-isothiocyanato, o-fluoro phenyl isothiocyanate, 2-fluorobenzenisothiocyanate, timtec-bb sbb006546
97520
1-fluoro-2-isothiocyanatobenzene
FC1=CC=CC=C1N=C=S
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-Fluorophenyl isothiocyanate
Densità 1.248
Punti di ebollizione 103°C to 104°C (12 mmHg)
Punto d'infiammabilità 97°C (206°F)
Indice di rifrazione 1.6254
Quantità 5 g
Numero UN UN2922
Beilstein 4176581
Sensibilità Moisture sensitive
Peso formulazione 153.18
Percent Purity 97%