missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylthiophene, 97%, Thermo Scientific™

Catalog Number 11392006
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1551-27-5
C7H10S
126.217
MFCD00014533
BTXIJTYYMLCUHI-UHFFFAOYSA-N
2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
73771
2-propylthiophene
CCCC1=CC=CS1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1551-27-5
C7H10S
126.217
MFCD00014533
BTXIJTYYMLCUHI-UHFFFAOYSA-N
2-n-propylthiophene, thiophene, 2-propyl, isopropylthiophene, 2-n-propyl thiophene, 2-propyl-thiophene, 2-propyl thiophene, thiophene, propyl, thiophene,2-propyl, 2-propylthiophene, 4-hydroxy-benzenesulfonamide
73771
2-propylthiophene
CCCC1=CC=CS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 2-n-Propylthiophene
Densità 0.984
Punti di ebollizione 158°C to 160°C
Punto d'infiammabilità 32°C (90°F)
Odore Characteristic
Indice di rifrazione 1.505
Quantità 5 g
Numero UN UN1993
Beilstein 105753
Peso formulazione 126.22
Percent Purity 97%
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'