missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitro-N-propylbenzamide, 97%, Thermo Scientific™

Catalog Number 15445518
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
10056-18-5
C10H11N3O5
253.214
MFCD01212114
LTJACLQPFWDXMR-UHFFFAOYSA-N
benzamide, 3,5-dinitro-n-propyl, 3,5-dinitro-n-propyl-benzamide, n-propyl-3,5-dinitrobenzamide
4417264
3,5-dinitro-N-propylbenzamide
CCCNC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
10056-18-5
C10H11N3O5
253.214
MFCD01212114
LTJACLQPFWDXMR-UHFFFAOYSA-N
benzamide, 3,5-dinitro-n-propyl, 3,5-dinitro-n-propyl-benzamide, n-propyl-3,5-dinitrobenzamide
4417264
3,5-dinitro-N-propylbenzamide
CCCNC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3,5-Dinitro-N-propylbenzamide
Quantità 250 mg
Peso formulazione 253.21
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'