missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,5-difluoropyridine, 97%, Thermo Scientific Chemicals

Catalog Number 15435387
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
851179-00-5
C5H2ClF2N
149.525
MFCD09998887
ATKDHGGWYZLPPJ-UHFFFAOYSA-N
3-chloro-2,5-difluoro-pyridine, pyridine, 3-chloro-2,5-difluoro, ksc654c8n
19371877
3-chloro-2,5-difluoropyridine
C1=C(C=NC(=C1Cl)F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
851179-00-5
C5H2ClF2N
149.525
MFCD09998887
ATKDHGGWYZLPPJ-UHFFFAOYSA-N
3-chloro-2,5-difluoro-pyridine, pyridine, 3-chloro-2,5-difluoro, ksc654c8n
19371877
3-chloro-2,5-difluoropyridine
C1=C(C=NC(=C1Cl)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Chloro-2,5-difluoropyridine
Indice di rifrazione 1.48
Quantità 1 g
Numero UN UN2924
Peso formulazione 149.53
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'