Learn More
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantità:
1 g
5 g
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Materiale o nome chimico | 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride |
Indice di rifrazione | 1.475 |
Quantità | 1 g |
Numero UN | UN3265 |
Sensibilità | Moisture sensitive |
Peso formulazione | 261 |
Percent Purity | 97% |