missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7030701
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-51-4
C9H7F3O3
220.147
MFCD08741402
OCLLNMRZSFBMBJ-UHFFFAOYSA-N
3-methyl-5-trifluoromethoxy benzoic acid, 5-methyl-3-trifluoromethoxy benzoic acid
26985612
3-methyl-5-(trifluoromethoxy)benzoic acid
CC1=CC(=CC(=C1)C(=O)O)OC(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
916420-51-4
C9H7F3O3
220.147
MFCD08741402
OCLLNMRZSFBMBJ-UHFFFAOYSA-N
3-methyl-5-trifluoromethoxy benzoic acid, 5-methyl-3-trifluoromethoxy benzoic acid
26985612
3-methyl-5-(trifluoromethoxy)benzoic acid
CC1=CC(=CC(=C1)C(=O)O)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 3-Methyl-5-(trifluoromethoxy)benzoic acid
Punto di fusione 81°C to 83°C
Quantità 1 g
Peso formulazione 220.15
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'