missing translation for 'onlineSavingsMsg'
Learn More

3M™ Gray Protective Coverall 4570

Codice prodotto. 15789027
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Dimensione della confezione:
Pezzo
pezzi 12
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15789027

Marca: 3M™ 7000089689

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Protective Coverall 4570 helps protect against a range of chemical jets and sprays, and has been certified to Type 3/4/5/6 levels of protection.

3M™ Protective Coverall 4570 offers chemical hold out and mechanical strength for workers in environments with chemical exposure. The suit helps protect against hazardous dusts (Type 5), light liquid splashes (Type 6), low pressure liquid sprays (Type 4) and high pressure liquid jets (Type 3). It is also certified to offer protection against radioactive particles (EN 1073-2) and (EN 14126). This suit achieves the highest class performance on all tests in EN 14126 and passes ASTM F1670 and F1671 for helping to protect against biological contaminants.

  • A Versatile Protective Suit for many Applications
  • Protective coverall helps protect against a range of chemical jets and sprays
  • Heavy-duty fabric with a soft anti-static treated inner layer offers protection and convenience
  • Stitched seams are sealed with a 5-layer chemically protective tape
  • Two storm flaps create a double seal for extra protection
  • Large thumb loops and zipper pulls offer convenience while wearing gloves
  • Knit polyester cuffs for user comfort
  • Anti-static, low-linting, microporous
  • Elastic around the head for secure fit
  • Elastic around the face to ensure secure fit
  • Elastic at the calf to provide closure around the top of the footwear for protection against contaminants
  • Elastic around the ankle for protection against contaminants and enhanced fit
  • Universal size with ties to provide a good fit for a wide range of users
  • Lightweight and comfortable to wear
TRUSTED_SUSTAINABILITY

Specifica

Size Large
Materiale Polyethylene
Colore Gray
Stile vita Elastic
Stile polso Elastic
Hood Type 3-panel
Tipo di chiusura Front Zipper with Double Storm Flap
Tipo di giuntura Taped
Senza lattice
Modello 4570-L
N. di tasche None
Misura petto (sistema imperiale) 39 – 43in.
Misura petto (sistema metrico) 100 to 108 cm
Da utilizzare con (applicazione) Chemical mixing and handling, chemical clean-up, hazardous waste remediation, decontamination, tank cleaning, handling toxic powders, infective agents
Altezza (sistema imperiale) 69 to 71 in.
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato