Learn More
3M™ Versaflo™TR-600 Series Filter Covers
Flter cover for use with TR-6320N, TR-6350N and TR-6360N cartridges
Marca: 3M™ 7100021336
13.65 EUR valido fino al 2024-12-31
Usa il codice promo "22912" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.20000kg
Descrizione
- Helps protect cartridge from physical damage
- Holds TR-6600 Prefilter or TR-662 Spark Arrestor in place
- Enables PAPR to be worn through decontamination showers or rain
- Easy to use
- 3M Catalog# TR-6300FC
Applications:
Recommended Industry: Healthcare, Pharmaceutical
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifica
Filter Cover TR-6300FC / 37380 (AAD) | |
TR-6300 Series Cartridges | |
TR-6300FC | |
1/Cs. |