Learn More
4-(4-Pyridyl)-2-thiazoleacetonitrile, 97%, Thermo Scientific™
Catalog Number p-7056379
Click to view available options
Quantità:
250 mg
1 g
1016888-78-0
C10H7N3S
201.247
MFCD09950233
LIIDMCZQVRSSQE-UHFFFAOYSA-N
4-4-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-4-yl thiazol-2-yl acetonitrile, 2-4-pyridin-4-yl-1,3-thiazol-2-yl acetonitrile
24710447
2-(4-pyridin-4-yl-1,3-thiazol-2-yl)acetonitrile
C1=CN=CC=C1C2=CSC(=N2)CC#N
1016888-78-0
C10H7N3S
201.247
MFCD09950233
LIIDMCZQVRSSQE-UHFFFAOYSA-N
4-4-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-4-yl thiazol-2-yl acetonitrile, 2-4-pyridin-4-yl-1,3-thiazol-2-yl acetonitrile
24710447
2-(4-pyridin-4-yl-1,3-thiazol-2-yl)acetonitrile
C1=CN=CC=C1C2=CSC(=N2)CC#N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-(4-Pyridyl)-2-thiazoleacetonitrile
Colore Yellow to Brown
Quantità 250 mg
Numero UN UN3439
Peso formulazione 201.25
Percent Purity 97%
Forma fisica Solid