missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Catalog Number 15447565
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
886500-93-2
C8H5BrClF3O
289.476
MFCD06660249
LAZINNAFCZCHLJ-UHFFFAOYSA-N
4-bromomethyl-1-chloro-2-trifluoromethoxy benzene, 4-chloro-3-trifluoromethoxy benzyl bromide, 4-chloro-3-trifluoromethoxybenzyl bromide, 2-chloro-5-bromomethyl-1-trifluoromethoxy benzene, benzene,4-bromomethyl-1-chloro-2-trifluoromethoxy
17750697
4-(bromomethyl)-1-chloro-2-(trifluoromethoxy)benzene
C1=CC(=C(C=C1CBr)OC(F)(F)F)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
886500-93-2
C8H5BrClF3O
289.476
MFCD06660249
LAZINNAFCZCHLJ-UHFFFAOYSA-N
4-bromomethyl-1-chloro-2-trifluoromethoxy benzene, 4-chloro-3-trifluoromethoxy benzyl bromide, 4-chloro-3-trifluoromethoxybenzyl bromide, 2-chloro-5-bromomethyl-1-trifluoromethoxy benzene, benzene,4-bromomethyl-1-chloro-2-trifluoromethoxy
17750697
4-(bromomethyl)-1-chloro-2-(trifluoromethoxy)benzene
C1=CC(=C(C=C1CBr)OC(F)(F)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Chloro-3-(trifluoromethoxy)benzyl bromide is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-3-(trifluoromethoxy)benzyl bromide
Quantità 250 mg
Numero UN UN3265
Informazioni di solubilità Not miscible or difficult to mix with water.
Peso formulazione 289.48
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'