missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-8-methoxy-2-methylquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 11430020
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
64951-58-2
C11H10ClNO
207.66
MFCD00272401
RHSCPZBPNUZBOA-UHFFFAOYSA-N
807583
4-chloro-8-methoxy-2-methylquinoline
COC1=C2N=C(C)C=C(Cl)C2=CC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
64951-58-2
C11H10ClNO
207.66
MFCD00272401
RHSCPZBPNUZBOA-UHFFFAOYSA-N
807583
4-chloro-8-methoxy-2-methylquinoline
COC1=C2N=C(C)C=C(Cl)C2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-8-methoxy-2-methylquinoline
Punto di fusione 89°C to 93°C
Quantità 1 g
Sinonimo 4-chloro-8-methoxyquinaldine, 4-chloro-8-methoxxyquinaldine, 4-chloro-8-methoxy-2-methyl-quinoline, quinoline,4-chloro-8-methoxy-2-methyl, acmc-20aiah, pubchem20521, cbmicro_008398, 4-chloro-2-methyl-8-methoxyquinoline, quinoline, 4-chloro-8-methoxy-2-methyl, 4-chloro-8-methoxy-2-methylquinoline
Peso formulazione 207.66
Percent Purity 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'