Learn More
4-Methyl-3,5-dinitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog Number 11404350
Click to view available options
Quantità:
1 g
5 g
25 g
28249-49-2
C7H7BN2O6
225.951
MFCD08060935
DTWIWVTWNMEZOL-UHFFFAOYSA-N
3,5-dinitro-4-methylbenzeneboronic acid, 4-methyl-3,5-dinitrophenyl boronic acid, 3,5-dinitro-4-methylphenylboronic acid, acmc-1cr7m, 3,5-dinitro-p-tolueneboronic acid, 3,5-dinitro-4-methylphenylboranic acid, 4-methyl-3,5-dinitrobenzeneboronic acid, p-tolueneboronic acid,3,5-dinitro-8ci, 3,5-dinitro-4-methylphenylboronic acid; 3,5-dinitro-p-tolueneboronic acid
17998947
(4-methyl-3,5-dinitrophenyl)boronic acid
B(C1=CC(=C(C(=C1)[N+](=O)[O-])C)[N+](=O)[O-])(O)O
28249-49-2
C7H7BN2O6
225.951
MFCD08060935
DTWIWVTWNMEZOL-UHFFFAOYSA-N
3,5-dinitro-4-methylbenzeneboronic acid, 4-methyl-3,5-dinitrophenyl boronic acid, 3,5-dinitro-4-methylphenylboronic acid, acmc-1cr7m, 3,5-dinitro-p-tolueneboronic acid, 3,5-dinitro-4-methylphenylboranic acid, 4-methyl-3,5-dinitrobenzeneboronic acid, p-tolueneboronic acid,3,5-dinitro-8ci, 3,5-dinitro-4-methylphenylboronic acid; 3,5-dinitro-p-tolueneboronic acid
17998947
(4-methyl-3,5-dinitrophenyl)boronic acid
B(C1=CC(=C(C(=C1)[N+](=O)[O-])C)[N+](=O)[O-])(O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Methyl-3,5-dinitrobenzeneboronic acid
Punto di fusione ∼280°C (decomposition)
Quantità 5 g
Peso formulazione 225.95
Percent Purity 97%