missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Propylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15479997
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
171922-46-6
C10H14BNO3
207.04
MFCD06656015
KWJCBWYHJKWCRD-UHFFFAOYSA-N
4-propylcarbamoyl phenyl boronic acid, 4-n-propylaminocarbonyl phenylboronic acid, 4-propylcarbamoyl phenylboronic acid, 4-propylaminocarbonyl benzeneboronic acid, 4-n-propylcarbamoyl benzeneboronic acid, n-propyl 4-boronobenzamide, acmc-1c1gn, 4-propylcarbamoyl phenyl boronicacid, 4-n-propylaminocarbonyl phenylboronicacid
2773569
[4-(propylcarbamoyl)phenyl]boronic acid
CCCNC(=O)C1=CC=C(C=C1)B(O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
171922-46-6
C10H14BNO3
207.04
MFCD06656015
KWJCBWYHJKWCRD-UHFFFAOYSA-N
4-propylcarbamoyl phenyl boronic acid, 4-n-propylaminocarbonyl phenylboronic acid, 4-propylcarbamoyl phenylboronic acid, 4-propylaminocarbonyl benzeneboronic acid, 4-n-propylcarbamoyl benzeneboronic acid, n-propyl 4-boronobenzamide, acmc-1c1gn, 4-propylcarbamoyl phenyl boronicacid, 4-n-propylaminocarbonyl phenylboronicacid
2773569
[4-(propylcarbamoyl)phenyl]boronic acid
CCCNC(=O)C1=CC=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-(n-Propylcarbamoyl)benzeneboronic acid
Punto di fusione 210°C to 214°C
Quantità 250 mg
Peso formulazione 207.04
Percent Purity 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'