missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzamide, 97%, Thermo Scientific™

Catalog Number 15446157
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1020113-65-8
C7H6F5NOS
247.183
MFCD16652448
MVRXMHQBQZQJMM-UHFFFAOYSA-N
4-pentafluorosulfur benzamide, 4-pentafluoro-??-sulfanyl benzamide, 4-pentafluorothio benzamide
59296134
4-(pentafluoro-$l^{6}-sulfanyl)benzamide
C1=CC(=CC=C1C(=O)N)S(F)(F)(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1020113-65-8
C7H6F5NOS
247.183
MFCD16652448
MVRXMHQBQZQJMM-UHFFFAOYSA-N
4-pentafluorosulfur benzamide, 4-pentafluoro-??-sulfanyl benzamide, 4-pentafluorothio benzamide
59296134
4-(pentafluoro-$l^{6}-sulfanyl)benzamide
C1=CC(=CC=C1C(=O)N)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-(Pentafluorothio)benzamide
Quantità 5 g
Peso formulazione 247.18
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'