missing translation for 'onlineSavingsMsg'
Learn More

5'-Chloro-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals

Catalog Number p-7022596
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1450-74-4
C8H7ClO2
170.592
MFCD00067788
XTGCUDZCCIRWHL-UHFFFAOYSA-N
5'-chloro-2'-hydroxyacetophenone, 1-5-chloro-2-hydroxyphenyl ethanone, 5-chloro-2-hydroxyacetophenone, 1-5-chloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4-chlorophenol, ethanone, 1-5-chloro-2-hydroxyphenyl, 2'-hydroxy-5'-chloroacetophenone, acetophenone, 5'-chloro-2'-hydroxy, 2-hydroxy-5-chloro acetophenone, 1-5-chloro-2-hydroxyphenyl-1-ethanone
74061
1-(5-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1450-74-4
C8H7ClO2
170.592
MFCD00067788
XTGCUDZCCIRWHL-UHFFFAOYSA-N
5'-chloro-2'-hydroxyacetophenone, 1-5-chloro-2-hydroxyphenyl ethanone, 5-chloro-2-hydroxyacetophenone, 1-5-chloro-2-hydroxyphenyl ethan-1-one, 2-acetyl-4-chlorophenol, ethanone, 1-5-chloro-2-hydroxyphenyl, 2'-hydroxy-5'-chloroacetophenone, acetophenone, 5'-chloro-2'-hydroxy, 2-hydroxy-5-chloro acetophenone, 1-5-chloro-2-hydroxyphenyl-1-ethanone
74061
1-(5-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 5'-Chloro-2'-hydroxyacetophenone
Punto di fusione 52°C to 53°C
Punti di ebollizione 107°C to 109°C (12 mmHg)
Quantità 1 g
Beilstein 1365589
Peso formulazione 170.6
Percent Purity 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'