Learn More
5-Cyclobutyl-5-phenylhydantoin, 97+%, Thermo Scientific™
Catalog Number p-7023061
Click to view available options
Quantità:
1 g
5 g
125650-44-4
C13H14N2O2
230.27
MFCD00022395
YFMSPNSDLBGLRF-UHFFFAOYNA-N
5-cyclobutyl-5-phenylhydantoin, acmc-20am3v, 5-cyclobutyl-5-phenyl-1,3-diazolidine-2,4-dione
277170
5-cyclobutyl-5-phenylimidazolidine-2,4-dione
O=C1NC(=O)C(N1)(C1CCC1)C1=CC=CC=C1
125650-44-4
C13H14N2O2
230.27
MFCD00022395
YFMSPNSDLBGLRF-UHFFFAOYNA-N
5-cyclobutyl-5-phenylhydantoin, acmc-20am3v, 5-cyclobutyl-5-phenyl-1,3-diazolidine-2,4-dione
277170
5-cyclobutyl-5-phenylimidazolidine-2,4-dione
O=C1NC(=O)C(N1)(C1CCC1)C1=CC=CC=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 5-Cyclobutyl-5-phenylhydantoin
Punto di fusione 234°C to 235°C
Quantità 1 g
Peso formulazione 230.27
Percent Purity ≥97%