missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-methylquinoline, 98%, Thermo Scientific Chemicals

Catalog Number 11403118
Click to view available options
Quantità:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
613-21-8
C10H9NO
159.188
MFCD00047600
USSQQASIZNTRAJ-UHFFFAOYSA-N
6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g
594395
2-methylquinolin-6-ol
CC1=NC2=C(C=C1)C=C(C=C2)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
613-21-8
C10H9NO
159.188
MFCD00047600
USSQQASIZNTRAJ-UHFFFAOYSA-N
6-hydroxy-2-methylquinoline, 6-hydroxyquinaldine, 2-methyl-6-quinolinol, 6-quinolinol, 2-methyl, 2-methyl-6-hydroxyquinoline, 2-methyl-quinolin-6-ol, acmc-1bit8, 2-methyl-6-quinolinol #, 6-hydroxy-2-methyl quionoline, 6-hydroxy-2-methylquinoline 1g
594395
2-methylquinolin-6-ol
CC1=NC2=C(C=C1)C=C(C=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 6-Hydroxy-2-methylquinoline
Quantità 5 g
Peso formulazione 159.19
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'