Learn More
Uvex™ 9192 Pheos S Spectacles
Duo-spherical lenses provide a wide field of vision with soft, non-slip arm ends to prevent pressure points for enhanced comfort. A narrow version of the original pheo series designed to provide an optimal, secure fit for those with smaller heads.
Marca:  Uvex™ 9192283
Dettagli aggiuntivi : Peso : 0.03700kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17237281
  • 15.90€ / Pezzo
Spedizione stimata: 26-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • A narrow version of the original pheos series to provide users with smaller heads an optimal, and secure fit
  • Soft, non-slip arm end prevents pressure points for enhanced comfort
  • Duo-spherical lenses provide a large field of vision
  • Design is extremely compatible with ear defenders, helmets, and bump caps
  • Eye Plus Protection provides additional eyebrow protection with a bevelled edge in the upper area
  • Easily add a headband and frame when working at heights or in confined spaces
Supravision Extreme Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant
  • Ideal for working areas with long-term exposure to condensation, and high levels of dirt
Supravision Excellence Coating
  • Interior: Anti-fog
  • Exterior: Scratch-resistant and chemical-resistant
  • Lenses are easy to clean and less susceptible to soiling
Specifica
Specifica
EN 166:2001, EN 172:1994 + A1:2000 + A2:2001
Gray
Black, Gray
0.23
Supravision Extreme
UV 400
Duo-Spherical
Working at Height or in Confined Spaces
Pheos S
No
Polycarbonate
Narrow