Learn More
Uvex 94900 Comfortable Climate Insole For Uvex G2 - Width 14
Comfortable climatic insole with antistatic properties for Uvex 1 G2
Marca: Uvex 9490048
Dettagli aggiuntivi : Peso : 0.06600kg
Descrizione
- Full-surface foot shock absorption
- Highly breathable, moisture-absorbent and quickly re-drying material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9490.0
- Sizes: 35 to 52
Specifica
Black | |
Microvelour | |
No | |
Wide width | |
No |
Uvex G2 | |
48 | |
Unisex | |
Yes |